Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032616.1 | NP_116005.1 | 9 | 1362 | ||
XM_006228844.2 | XP_006228906.1 | 8 | 1329 | |||
XM_006228845.2 | XP_006228907.1 | 7 | 1182 | |||
GenBank mRNA | AF119667.1 | - | 9 | 1362 | ||
AF119668.1 | - | 8 | 1305 | |||
AF119669.1 | - | 7 | 1158 | |||
BC100626.1 | - | 8 | 1302 | 7936432 |