Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022620.1 | NP_072142.1 | 7 | 889 | ||
XM_008760793.1 | XP_008759015.1 | 8 | 1132 | |||
GenBank mRNA | AF102129.1 | - | 7 | 889 | ||
BC097318.1 | - | 8 | 1044 |
Gene Symbol: LOC103691498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760834.1 | XP_008759056.1 | 4 | 544 |