Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phka1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022626.1 | NP_072148.1 | 18 | 1951 | ||
XM_006257101.2 | XP_006257163.1 | 18 | 2278 | |||
XM_006257102.2 | XP_006257164.1 | 18 | 2278 | |||
GenBank mRNA | AF197561.1 | - | 18 | 1951 | ||
M92918.1 | - | 3 | 313 |