Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022960.2 | NP_075249.1 | 8 | 918 | ||
XM_006243362.1 | XP_006243424.1 | 7 | 1023 | |||
XM_008766336.1 | XP_008764558.1 | 4 | 724 | |||
GenBank mRNA | AF016406.1 | - | 4 | 697 | ||
BC085731.1 | - | 8 | 918 | 7134715 | ||
FQ209455.1 | - | 4 | 719 | |||
FQ211101.1 | - | 4 | 639 | |||
FQ212527.1 | - | 4 | 602 | |||
FQ214432.1 | - | 7 | 1028 | |||
FQ218988.1 | - | 4 | 722 | |||
FQ219175.1 | - | 4 | 639 |