Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Arhgef9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023957.1 | NP_076447.1 | 5 | 1161 | ||
GenBank mRNA | AJ250425.1 | - | 5 | 1161 | ||
AJ302676.1 | - | 4 | 981 |