Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Scn9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133289.1 | NP_579823.1 | 26 | 5709 | ||
GenBank mRNA | AF000368.1 | - | 26 | 5709 | ||
U79568.1 | - | 26 | 5709 |