Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pggt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031082.1 | NP_112344.1 | 9 | 990 | ||
XM_006254691.2 | XP_006254753.1 | 9 | 2230 | |||
XM_008772177.1 | XP_008770399.1 | 10 | 992 | |||
GenBank mRNA | L24116.1 | - | 9 | 990 |