Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031337.2 | NP_112627.2 | 8 | 1608 | ||
XM_006236676.1 | XP_006236738.1 | 7 | 1269 | |||
XM_006236677.1 | XP_006236739.1 | 8 | 1411 | |||
XM_006236679.2 | XP_006236741.1 | 7 | 1402 | |||
XM_006236680.1 | XP_006236742.1 | 7 | 1285 | |||
XM_006236682.1 | XP_006236744.1 | 6 | 1096 | |||
XM_006236684.2 | XP_006236746.1 | 6 | 1290 | |||
XM_008762996.1 | XP_008761218.1 | 7 | 1431 | |||
XM_008762997.1 | XP_008761219.1 | 6 | 1319 | |||
GenBank mRNA | AB018049.1 | - | 7 | 1274 | ||
BC078798.1 | - | 8 | 1608 | 7106217 |