Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031355.1 | NP_112645.1 | 4 | 344 | ||
XM_006253358.2 | XP_006253420.1 | 5 | 347 | |||
GenBank mRNA | AB039664.1 | - | 4 | 344 | ||
AF048829.1 | - | 3 | 228 | |||
AF048830.1 | - | 3 | 231 | |||
AF268469.1 | - | 3 | 255 | |||
BC061780.1 | - | 4 | 280 | 5623339 |