Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031825.1 | NP_114013.1 | 45 | 5567 | ||
XM_006234935.2 | XP_006234997.1 | 46 | 5856 | |||
XM_008762202.1 | XP_008760424.1 | 45 | 5757 | |||
XM_008762203.1 | XP_008760425.1 | 46 | 5851 | |||
GenBank mRNA | AF135059.1 | - | 45 | 5567 |