Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031828.1 | NP_114016.1 | 3 | 1061 | ||
GenBank mRNA | AF083341.1 | - | 3 | 497 | ||
AF135265.1 | - | 3 | 915 | |||
AY330290.1 | - | 3 | 386 | |||
AY330291.1 | - | 3 | 386 | |||
AY330292.1 | - | 3 | 386 | |||
AY330293.2 | - | 3 | 386 | |||
AY330294.1 | - | 3 | 386 | |||
AY344964.1 | - | 3 | 386 | |||
AY344965.2 | - | 3 | 890 | |||
U40603.1 | - | 3 | 1061 | |||
U55995.1 | - | 3 | 539 | |||
U93052.1 | - | 3 | 487 |