Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Src | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031977.1 | NP_114183.1 | 11 | 1386 | ||
XM_008762387.1 | XP_008760609.1 | 11 | 1395 | |||
XM_008762388.1 | XP_008760610.1 | 11 | 1432 | |||
XM_008762389.1 | XP_008760611.1 | 11 | 1503 | |||
GenBank mRNA | AF130457.1 | - | 10 | 1368 | ||
AF157016.1 | - | 11 | 1386 | |||
DQ120509.1 | - | 10 | 1352 | |||
DQ120510.1 | - | 10 | 1352 |