Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fetub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053348.1 | NP_445800.1 | 6 | 822 | ||
XM_006248549.1 | XP_006248611.1 | 7 | 776 | |||
XM_006248550.1 | XP_006248612.1 | 7 | 1139 | |||
GenBank mRNA | AJ242926.1 | - | 6 | 822 | ||
BC070508.1 | - | 7 | 761 | 7099154 | ||
FQ209398.1 | - | 6 | 834 | |||
FQ209518.1 | - | 6 | 773 | |||
FQ209610.1 | - | 6 | 809 | |||
FQ209832.1 | - | 6 | 774 | |||
FQ210335.1 | - | 6 | 774 | |||
FQ211050.1 | - | 6 | 773 | |||
FQ218476.1 | - | 6 | 804 | |||
FQ218884.1 | - | 7 | 744 | |||
FQ218893.1 | - | 5 | 674 | |||
FQ218930.1 | - | 6 | 806 | |||
FQ218949.1 | - | 6 | 623 | |||
FQ219048.1 | - | 6 | 804 | |||
FQ219152.1 | - | 6 | 810 | |||
FQ219190.1 | - | 6 | 809 | |||
FQ219266.1 | - | 6 | 773 | |||
FQ219352.1 | - | 6 | 773 | |||
FQ219426.1 | - | 4 | 375 | |||
FQ219454.1 | - | 6 | 773 | |||
FQ219459.1 | - | 6 | 771 | |||
FQ219500.1 | - | 6 | 776 | |||
FQ219513.1 | - | 6 | 845 |