Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cdh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053393.2 | NP_445845.2 | 10 | 2097 | ||
XM_008772244.1 | XP_008770466.1 | 9 | 1444 | |||
GenBank mRNA | AB010436.1 | - | 10 | 2097 |