Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Picalm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053554.2 | NP_446006.1 | 4 | 408 | ||
XM_006229669.2 | XP_006229731.1 | 4 | 678 | |||
XM_006229670.2 | XP_006229732.1 | 4 | 678 | |||
XM_006229671.2 | XP_006229733.1 | 4 | 679 | |||
XM_006229672.2 | XP_006229734.1 | 4 | 678 | |||
XM_006229673.2 | XP_006229735.1 | 4 | 678 | |||
XM_006229674.2 | XP_006229736.1 | 4 | 678 | |||
XM_006229675.2 | XP_006229737.1 | 4 | 678 | |||
XM_006229676.2 | XP_006229738.1 | 4 | 677 | |||
XM_006229677.2 | XP_006229739.1 | 4 | 677 | |||
XM_006229678.2 | XP_006229740.1 | 4 | 677 | |||
XM_006229679.2 | XP_006229741.1 | 4 | 677 | |||
XM_006229680.2 | XP_006229742.1 | 4 | 677 | |||
XM_006229681.2 | XP_006229743.1 | 4 | 677 | |||
XM_006229682.2 | XP_006229744.1 | 4 | 677 | |||
XM_006229683.2 | XP_006229745.1 | 4 | 677 | |||
XM_006229684.2 | XP_006229746.1 | 4 | 677 | |||
XM_006229685.2 | XP_006229747.1 | 4 | 677 | |||
GenBank mRNA | AF041373.1 | - | 4 | 409 | ||
AF041374.1 | - | 4 | 409 | |||
AY699717.1 | - | 4 | 384 | |||
AY706215.1 | - | 4 | 384 | |||
AY724477.1 | - | 4 | 402 | |||
BC100142.1 | - | 4 | 448 | 7456159 |