Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sep15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133297.2 | NP_579831.2 | 3, 4 | 573 | ||
GenBank mRNA | AF390544.1 | - | 3, 4 | 297 | ||
BC060547.1 | - | 3, 4 | 322 | 6921948 | ||
CK596377.1 | - | 3, 4 | 588 | |||
DY561321.1 | - | 3, 4 | 311 | |||
FQ209574.1 | - | 3, 4 | 308 | |||
FQ212727.1 | - | 3, 4 | 341 | |||
FQ213177.1 | - | 3, 4 | 322 | |||
FQ213838.1 | - | 3, 4 | 308 | |||
FQ214634.1 | - | 3, 4 | 318 | |||
FQ215362.1 | - | 3, 4 | 308 | |||
FQ215992.1 | - | 3, 4 | 331 | |||
FQ216831.1 | - | 3, 4 | 308 | |||
FQ219237.1 | - | 3, 4 | 329 | |||
FQ219570.1 | - | 3, 4 | 308 | |||
FQ219725.1 | - | 3, 4 | 301 | |||
FQ220311.1 | - | 3, 4 | 307 | |||
FQ220593.1 | - | 3, 4 | 313 | |||
FQ229133.1 | - | 3, 4 | 314 | |||
FQ230131.1 | - | 3, 4 | 308 |