Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WNT7A SIRNA Assays ›
Gene Symbol:
Wnt7a
Gene Name:
wingless-type MMTV integration site family, member 7A
Gene Aliases:
-
Chromosome Location:
Chr.4: 122994425 - 123040609 on Build Rnor_6.0
UniGene ID:
Rn.230035
Species:
Rat
siRNA ID
RSS331929
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Wnt7a
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100473.1
NP_001093943.1
2
472
Back To Top
More Information
Panther Classification:
Molecular Function -
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
embryonic axis specification
cartilage condensation
angiogenesis
chondrocyte differentiation
neurotransmitter secretion
asymmetric protein localization
positive regulation of cell proliferation
dorsal/ventral pattern formation
positive regulation of endothelial cell migration
positive regulation of gene expression
skeletal muscle satellite cell activation
skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
cerebellar granule cell differentiation
cell proliferation in forebrain
central nervous system vasculogenesis
establishment of cell polarity
neuron differentiation
embryonic limb morphogenesis
regulation of axon diameter
response to estradiol
somatic stem cell population maintenance
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
forelimb morphogenesis
hindlimb morphogenesis
non-canonical Wnt signaling pathway
Wnt signaling pathway involved in wound healing, spreading of epidermal cells
wound healing
regulation of cell proliferation
embryonic digit morphogenesis
negative regulation of apoptotic process
skin morphogenesis
response to estrogen
cell fate commitment
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
somatic stem cell division
reproductive structure development
skeletal system morphogenesis
stem cell development
negative regulation of neurogenesis
regulation of axonogenesis
synapse organization
cartilage development
positive regulation of synapse assembly
palate development
positive regulation of epithelial cell proliferation involved in wound healing
uterus development
oviduct development
canonical Wnt signaling pathway
limb development
uterus morphogenesis
cellular response to transforming growth factor beta stimulus
positive regulation of canonical Wnt signaling pathway
receptor binding
frizzled binding
cytokine activity
protein binding
receptor agonist activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-lzswf:80/10.239.160.35:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0