Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacsin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130740.2 | NP_570096.2 | 11 | 1650 | ||
XM_006242049.2 | XP_006242111.1 | 10 | 1868 | |||
XM_006242050.2 | XP_006242112.1 | 12 | 2349 | |||
XM_006242051.1 | XP_006242113.1 | 12 | 1598 | |||
XM_006242052.2 | XP_006242114.1 | 12 | 2259 | |||
XM_006242053.1 | XP_006242115.1 | 12 | 1621 | |||
XM_006242054.2 | XP_006242116.1 | 9 | 1745 | |||
XM_006242055.1 | XP_006242117.1 | 11 | 1592 | |||
XM_006242057.2 | XP_006242119.1 | 11 | 2227 | |||
XM_006242058.1 | XP_006242120.1 | 11 | 1475 | |||
XM_006242059.2 | XP_006242121.1 | 10 | 2220 | |||
XM_006242060.1 | XP_006242122.1 | 10 | 1469 | |||
XM_008765660.1 | XP_008763882.1 | 11 | 6097 | |||
XM_008765661.1 | XP_008763883.1 | 11 | 2216 | |||
XM_008765662.1 | XP_008763884.1 | 11 | 1456 | |||
GenBank mRNA | AF139492.1 | - | 12 | 1574 | ||
AF139493.1 | - | 11 | 1451 | |||
AF139494.1 | - | 11 | 1568 | |||
AF139495.1 | - | 10 | 1445 | |||
BC070911.1 | - | 11 | 1650 | 7104542 | ||
FQ230278.1 | - | 4 | 418 |