- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Trim63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080903.1 | NP_543179.1 | 3 | 522 | ||
GenBank mRNA | AY059627.1 | - | 3 | 522 | ||
BC061824.1 | - | 3 | 509 | 5599072 |