Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt1a7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130407.2 | NP_569091.2 | 758 | |||
GenBank mRNA | AF461738.1 | - | 766 | |||
AY435134.1 | - | 725 | ||||
BC078732.1 | - | 761 | ||||
U75903.1 | - | 767 |