Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Folr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133527.1 | NP_598211.1 | 4 | 406 | ||
XM_006229842.2 | XP_006229904.1 | 4 | 527 | |||
GenBank mRNA | AF219904.1 | - | 4 | 406 | ||
BC157811.1 | - | 3 | 389 |