Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd79b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133533.2 | NP_598217.1 | 3, 4 | 475 | ||
GenBank mRNA | BC088115.1 | - | 3, 4 | 475 | 7325406 | |
FQ226266.1 | - | 3, 4 | 472 | |||
FQ227061.1 | - | 3, 4 | 470 | |||
FQ227788.1 | - | 3, 4 | 464 | |||
FQ230261.1 | - | 3, 4 | 466 | |||
FQ232054.1 | - | 3, 4 | 466 | |||
FQ232680.1 | - | 3, 4 | 466 | |||
FQ232790.1 | - | 3, 4 | 489 | |||
FQ233507.1 | - | 3, 4 | 464 | |||
FQ234082.1 | - | 3, 4 | 449 | |||
FQ234487.1 | - | 3, 4 | 464 |