Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012560.2 | NP_036692.1 | 1 | 1383 | ||
GenBank mRNA | M87634.1 | - | 1 | 1384 |