Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139255.3 | NP_640348.2 | 8 | 1196 | ||
GenBank mRNA | AY095481.1 | - | 2 | 228 | ||
BC086408.1 | - | 3 | 329 | |||
BC097934.1 | - | 4 | 502 | |||
BC104715.1 | - | 3 | 267 | |||
BC126078.1 | - | 4 | 400 | |||
BC161835.1 | - | 4 | 358 |