Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017058.1 | NP_058754.1 | 8 | 919 | ||
GenBank mRNA | J04147.1 | - | 8 | 919 |