Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC24906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031537.1 | NP_113725.1 | 6 | 646 | ||
XM_006246376.2 | XP_006246438.1 | 6 | 696 | |||
XM_006246377.2 | XP_006246439.1 | 6 | 726 | |||
GenBank mRNA | AF041083.1 | - | 6 | 646 |