Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024353.1 | NP_077329.1 | 21 | 1955 | ||
XM_006235083.2 | XP_006235145.1 | 22 | 2025 | |||
XM_006235085.2 | XP_006235147.1 | 22 | 1886 | |||
XM_006235088.2 | XP_006235150.1 | 21 | 4139 | |||
XM_006235089.2 | XP_006235151.1 | 22 | 4712 | |||
XM_006235094.2 | XP_006235156.1 | 21 | 1989 | |||
XM_008762248.1 | XP_008760470.1 | 22 | 4258 | |||
XM_008762249.1 | XP_008760471.1 | 21 | 4222 | |||
XM_008762250.1 | XP_008760472.1 | 22 | 4258 | |||
XM_008762251.1 | XP_008760473.1 | 21 | 4222 | |||
GenBank mRNA | AF027571.1 | - | 4 | 344 | ||
AF031370.1 | - | 19 | 1722 | |||
L15556.1 | - | 21 | 1955 | |||
L18962.1 | - | 21 | 1855 | |||
U57836.1 | - | 21 | 1855 |