Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177593.1 | NP_001171064.1 | 13 | 1650 | ||
NM_013088.2 | NP_037220.2 | 13 | 1638 | |||
GenBank mRNA | D83016.1 | - | 7 | 687 | ||
U05963.1 | - | 13 | 1581 | |||
U09307.1 | - | 13 | 1563 |