Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000052.1 | NP_001000052.1 | 1 | 886 |
Gene Symbol: LOC103693566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768681.1 | XP_008766903.1 | 1 | 886 |