Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wbscr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025112.1 | NP_001020283.1 | 7, 8 | 1268 | ||
XM_008769132.1 | XP_008767354.1 | 7, 8 | 1136 | |||
GenBank mRNA | AB040671.1 | - | 7, 8 | 1268 |