Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106003.1 | NP_001099473.1 | 5 | 682 | ||
XM_006250895.2 | XP_006250957.1 | 6 | 753 |