Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prorsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106022.1 | NP_001099492.1 | 1, 2 | 126 | ||
GenBank mRNA | BC167083.1 | - | 1, 2 | 88 | 5620957 |