Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cav3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019155.2 | NP_062028.1 | 1 | 231 | ||
GenBank mRNA | BC084692.1 | - | 1 | 231 | 7103841 | |
U31968.1 | - | 1 | 229 |