Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106228.1 | NP_001099698.1 | 6 | 669 | ||
XM_006228395.2 | XP_006228457.1 | 6 | 687 | |||
XM_006228396.1 | XP_006228458.1 | 6 | 647 | |||
GenBank mRNA | FQ225470.1 | - | 6 | 658 |