Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019186.1 | NP_062059.1 | 2 | 341 | ||
XM_006240051.2 | XP_006240113.1 | 2 | 428 | |||
XM_006240052.2 | XP_006240114.1 | 2 | 477 | |||
GenBank mRNA | BC088148.1 | - | 2 | 427 | 7373646 | |
X77235.1 | - | 2 | 341 |