Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001079250.5 | XP_001079250.1 | 1 | 934 | ||
XM_006230199.2 | XP_006230261.1 | 2 | 819 |