Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170573.1 | NP_001164044.1 | 9, 10 | 913 | ||
GenBank mRNA | BC168887.1 | - | 7, 8 | 506 | ||
FQ229851.1 | - | 9, 10 | 848 |
Gene Symbol: LOC100910130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749779.3 | XP_003749827.1 | 9, 10 | 897 |