Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnni1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017184.1 | NP_058880.1 | 5 | 287 | ||
XM_006249862.2 | XP_006249924.1 | 4 | 1794 | |||
XM_006249864.1 | XP_006249926.1 | 4 | 373 | |||
XM_006249865.2 | XP_006249927.1 | 7 | 416 | |||
XM_006249866.2 | XP_006249928.1 | 7 | 347 | |||
XM_006249868.2 | XP_006249930.1 | 5 | 382 | |||
XM_006249869.2 | XP_006249931.1 | 5 | 306 | |||
GenBank mRNA | FQ214889.1 | - | 7 | 394 | ||
FQ215520.1 | - | 7 | 369 | |||
FQ215574.1 | - | 7 | 367 | |||
FQ215749.1 | - | 7 | 367 | |||
FQ216094.1 | - | 7 | 367 | |||
FQ216822.1 | - | 7 | 367 | |||
J04993.1 | - | 5 | 287 |