Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou3f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172085.1 | NP_742082.1 | 1 | 911 | ||
GenBank mRNA | L27663.1 | - | 1 | 911 |