Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ell3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011957.1 | NP_001011957.1 | 9 | 961 | ||
XM_006234831.2 | XP_006234893.1 | 9 | 993 | |||
XM_006234832.2 | XP_006234894.1 | 9 | 848 | |||
GenBank mRNA | BC084693.1 | - | 9 | 961 | 7107022 |