Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106691.1 | NP_001100161.1 | 1 | 429 | ||
XM_006239150.2 | XP_006239212.1 | 4 | 1420 | |||
XM_006239152.2 | XP_006239214.1 | 3 | 1365 | |||
XM_008764217.1 | XP_008762439.1 | 3 | 780 | |||
XM_008764218.1 | XP_008762440.1 | 4 | 688 | |||
XM_008764219.1 | XP_008762441.1 | 3 | 896 |