Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037775.2 | NP_001032864.1 | 1 | 554 | ||
XM_006248935.1 | XP_006248997.1 | 2 | 648 | |||
XM_006248936.2 | XP_006248998.1 | 2 | 1697 | |||
XM_006248937.2 | XP_006248999.1 | 2 | 2435 | |||
XM_008768971.1 | XP_008767193.1 | 3 | 1579 | |||
GenBank mRNA | BC100096.1 | - | 1 | 558 | 7375118 |