Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LOC304396 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013981.1 | NP_001014003.1 | 5, 6 | 695 | ||
XM_006249156.2 | XP_006249218.1 | 5, 6 | 781 | |||
GenBank mRNA | BC081940.1 | - | 5, 6 | 695 | 7115744 |