Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aftph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251532.2 | XP_006251594.1 | 7 | 2641 | ||
XM_006251534.2 | XP_006251596.1 | 7 | 2478 | |||
XM_008770443.1 | XP_008768665.1 | 8 | 2795 | |||
XM_223651.8 | XP_223651.5 | 7 | 2640 | |||
GenBank mRNA | FQ230354.1 | - | 6 | 831 |