Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107778.1 | NP_001101248.1 | 8 | 894 | ||
XM_006235027.2 | XP_006235089.1 | 9 | 1049 | |||
XM_006235028.2 | XP_006235090.1 | 8 | 929 | |||
XM_008762178.1 | XP_008760400.1 | 8 | 1970 |