Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107890.1 | NP_001101360.1 | 3, 4 | 696 | ||
XM_006237494.2 | XP_006237556.1 | 7, 8 | 827 | |||
XM_006237495.2 | XP_006237557.1 | 6, 7 | 643 | |||
XM_006237496.2 | XP_006237558.1 | 6, 7 | 706 | |||
XM_006237498.2 | XP_006237560.1 | 5, 6 | 526 |