Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007001.1 | NP_001007002.1 | 17 | 1575 | ||
XM_006238398.2 | XP_006238460.1 | 17 | 1683 | |||
GenBank mRNA | BC083611.1 | - | 17 | 1575 | 7133514 |