Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009601.1 | NP_001009601.1 | 6 | 927 | ||
XM_006244516.2 | XP_006244578.1 | 6 | 794 | |||
XM_006244517.2 | XP_006244579.1 | 6 | 1146 | |||
GenBank mRNA | BC087116.1 | - | 6 | 927 | 7193915 |