Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001003.1 | NP_001001003.1 | 1 | 749 | ||
XM_006251847.2 | XP_006251909.1 | 2 | 778 |
Set Membership: |
G-protein coupled receptor |