Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021659.1 | NP_067691.1 | 5 | 764 | ||
XM_006231065.2 | XP_006231127.1 | 9 | 1759 | |||
XM_006231066.2 | XP_006231128.1 | 8 | 2155 | |||
XM_006231067.2 | XP_006231129.1 | 8 | 1627 | |||
XM_006231068.2 | XP_006231130.1 | 8 | 1531 | |||
XM_006231069.2 | XP_006231131.1 | 6 | 1039 | |||
GenBank mRNA | AF336852.1 | - | 10 | 1579 | ||
AF336853.1 | - | 9 | 1351 | |||
AF336854.1 | - | 5 | 604 | |||
AF336855.1 | - | 9 | 1456 | |||
AF336856.1 | - | 7 | 955 | |||
AF336857.1 | - | 7 | 964 | |||
AF336858.1 | - | 7 | 1096 | |||
AF336859.1 | - | 8 | 1228 | |||
AF336860.1 | - | 8 | 1324 | |||
U20106.1 | - | 5 | 764 |